Skip to Content
Merck
All Photos(1)

Key Documents

733652

Sigma-Aldrich

Hexaethylene glycol di-p-toluenesulfonate

>97%

Synonym(s):

Polyethylene glycol, HEG ditosylate, Hexaethylene glycol ditosylate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€61.30

€61.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€61.30

About This Item

Empirical Formula (Hill Notation):
C26H38O11S2
CAS Number:
Molecular Weight:
590.70
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€61.30


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

>97%

form

liquid

refractive index

n20/D 1.521

density

1.227 g/mL at 25 °C

SMILES string

Cc1ccc(cc1)S(=O)(=O)OCCOCCOCCOCCOCCOCCOS(=O)(=O)c2ccc(C)cc2

InChI

1S/C26H38O11S2/c1-23-3-7-25(8-4-23)38(27,28)36-21-19-34-17-15-32-13-11-31-12-14-33-16-18-35-20-22-37-39(29,30)26-9-5-24(2)6-10-26/h3-10H,11-22H2,1-2H3

InChI key

ZIZXHPCBPDNLDD-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service