Skip to Content
Merck
All Photos(1)

Key Documents

339024

Sigma-Aldrich

Diethylene glycol di(p-toluenesulfonate)

98%

Synonym(s):

2,2′-Oxydiethyl ditosylate, Bis(2-tosyloxyethyl)ether, Diethylene glycol ditosylate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€109.00

€109.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€109.00

About This Item

Linear Formula:
(CH3C6H4SO3CH2CH2)2O
CAS Number:
Molecular Weight:
414.49
Beilstein:
583800
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€109.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

mp

87-89 °C (lit.)

functional group

ether
tosylate

SMILES string

Cc1ccc(cc1)S(=O)(=O)OCCOCCOS(=O)(=O)c2ccc(C)cc2

InChI

1S/C18H22O7S2/c1-15-3-7-17(8-4-15)26(19,20)24-13-11-23-12-14-25-27(21,22)18-9-5-16(2)6-10-18/h3-10H,11-14H2,1-2H3

InChI key

VYVPNTJBGPQTFA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diethylene glycol di(p-toluenesulfonate) was used in the synthesis of bis(dibenzosulfoxo)-27,30,35,38-tetramethyl-24-crown-6,[1] bis(dibenzosulfoxo)-21,24,29,32-tetramethyl-18-crown-4,[1] dibenzosulfoxo-15,18-dimethyl-12-crown-3[1] and dibenzosulfoxo-12,15-dimethyl-9-crown-2 macrocycles.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of new benzosulfoxide macrocyclic compounds.
Shockravi A, et al.
Phosphorus, Sulfur, and Silicon and the Related Elements, 179(3), 535-541 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service