Skip to Content
Merck
All Photos(1)

Documents

730599

Sigma-Aldrich

Cycloheptylmagnesium bromide solution

2 M in diethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13BrMg
Molecular Weight:
201.39
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solution

reaction suitability

reaction type: Grignard Reaction

concentration

2 M in diethyl ether

density

1.034 g/mL at 25 °C

SMILES string

Br[Mg]C1CCCCCC1

InChI

1S/C7H13.BrH.Mg/c1-2-4-6-7-5-3-1;;/h1H,2-7H2;1H;/q;;+1/p-1

InChI key

KMAQJJKRGAFLBH-UHFFFAOYSA-M

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service