Skip to Content
Merck
All Photos(1)

Key Documents

687952

Sigma-Aldrich

(Cyclohexylmethyl)magnesium bromide solution

Synonym(s):

Cyclohexylmethylmagnesium bromide

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€266.00

€266.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
€266.00

About This Item

Empirical Formula (Hill Notation):
C7H13BrMg
CAS Number:
Molecular Weight:
201.39
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€266.00


Check Cart for Availability

Request a Bulk Order

form

liquid

reaction suitability

reaction type: Grignard Reaction

density

0.966 g/mL at 25 °C

SMILES string

Br[Mg]CC1CCCCC1

InChI

1S/C7H13.BrH.Mg/c1-7-5-3-2-4-6-7;;/h7H,1-6H2;1H;/q;;+1/p-1

InChI key

ZBCIOGFGZRNNEC-UHFFFAOYSA-M

Application

Reactant involved in:
  • Isomerization polymerization of alkenylcyclohexanes
  • Intramolecular rearrangements of vinylidenecyclopropanes
  • Cycloisomerization of cyclic and acyclic enynes

Additionally reactant involved in synthesis of biologically active molecules including:
  • Dipeptidyl boronic acid proteasome inhibitors
  • Substituted 1,3-dihydroindole-2-ones with antitumor effects
  • Sulfur-free transition state nucleoside analog inhibitors of methylthioadenosine nucleosidase and phosphorylase

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 °F

Flash Point(C)

-20 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What solution is this in (ie: THF)? What is the concentration (ie: 0.2M, 1M, 2M...)?

    1 answer
    1. The concentration of this product ranges from 0.40 - 0.60 M in THF. The exact concentration is listed on the lot-specific Certificate of Analysis. Please see the sample Certificate of Analysis: https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/687952-50ML0000235108.pdf

      Please see the link below to the SDS which lists each component in Section 3:
      https://www.sigmaaldrich.com/sds/aldrich/687952

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service