Skip to Content
Merck
All Photos(1)

Key Documents

723126

Sigma-Aldrich

Bis(dodecylsulfanylthiocarbonyl) disulfide

≥95%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€442.00

€442.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€442.00

About This Item

Empirical Formula (Hill Notation):
C26H50S6
CAS Number:
Molecular Weight:
555.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.23

€442.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

solid

mp

30-35 °C

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCSC(=S)SSC(=S)SCCCCCCCCCCCC

InChI

1S/C26H50S6/c1-3-5-7-9-11-13-15-17-19-21-23-29-25(27)31-32-26(28)30-24-22-20-18-16-14-12-10-8-6-4-2/h3-24H2,1-2H3

InChI key

UUNRYKCXJSDLRD-UHFFFAOYSA-N

General description

Need help choosing the correct RAFT Agent? Please consult the RAFT Agent to Monomer compatibility table.

Application

Precursor for the synthesis of RAFT agents for controlled radical polymerization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RAFT Agent Design and Synthesis
Keddie, D. J.; et al.
Macromolecules, 45, 5321-5342 (2012)
Massimo Benaglia et al.
Journal of the American Chemical Society, 131(20), 6914-6915 (2009-05-01)
The polymerization of most monomers that are polymerizable by radical polymerization can be controlled by the reversible addition-fragmentation chain transfer (RAFT) process. However, it is usually required that the RAFT agent be selected according to the types of monomer being

Articles

RAFT polymerization uses commercial agents to control polymer properties without cytotoxic heavy metals like ATRP.

RAFT polymerization offers living characteristics to radical polymerization, contributing versatility to reversible deactivation radical polymerization methods.

Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.

Protocols

RAFT polymerization offers precise control, enabling tailored synthesis of complex polymer structures.

We presents an article featuring procedures that describe polymerization of methyl methacrylate and vinyl acetate homopolymers and a block copolymer as performed by researchers at CSIRO.

We present an article about RAFT, or Reversible Addition/Fragmentation Chain Transfer, which is a form of living radical polymerization.

Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service