Skip to Content
Merck
All Photos(2)

Key Documents

173029

Sigma-Aldrich

Bis(carboxymethyl)trithiocarbonate

98%

Synonym(s):

Bis(carboxymethyl) trithiocarbonate, Di(carboxymethyl) trithiocarbonate, Trithiocarbodiglycolic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€147.00

€147.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€147.00

About This Item

Linear Formula:
(HO2CCH2S)2CS
CAS Number:
Molecular Weight:
226.29
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€147.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

solid

mp

172-175 °C (lit.)

SMILES string

OC(=O)CSC(=S)SCC(O)=O

InChI

1S/C5H6O4S3/c6-3(7)1-11-5(10)12-2-4(8)9/h1-2H2,(H,6,7)(H,8,9)

InChI key

GQECANUIPBFPLA-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huazhen Tao et al.
Polymers, 11(5) (2019-05-18)
Photocatalyzed polymerization using organic molecules as catalysts has attracted broad interest because of its easy operation in ambient environments and low toxicity compared with metallic catalysts. In this work, we reported that 4,7-di(thiophen-2-yl)benzo[c][1,2,5]thiadiazole (DTBT) can act as an efficient photoredox

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service