Skip to Content
Merck
All Photos(1)

Key Documents

719102

Sigma-Aldrich

Organic conductive inks kit

Synonym(s):

OC ink kit, OC ink system, Plexcore® OC ink system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103

form

liquid

storage temp.

2-8°C

General description

Component 1: Plexcore® OC RG-1110 organic conductive ink
Form: liquid
Color: Dark blue
Resistivity: 10 to 300 ohm-cm
Viscosity: 5 - 11 cP
pH: 2.5 - 4.5
Surface Tension: 33-40 dyne/cm
Work Function: -4.8 eV (Scanning Kelvin Probe)
Density: ~0.95 g/cm3


Component 2: Plexcore® OC RG-1115 organic conductive ink
Form: liquid
Color: Dark blue
Resistivity: 10 to 300 ohm-cm
Viscosity: 5 - 11 cP
pH: 2.5 - 4.5
Surface Tension: 33-40 dyne/cm
Work Function: -5.25 eV (Scanning Kelvin Probe)
Density: ~0.95 g/cm3


Component 3: Plexcore® OC RG-1150 organic conductive ink
Form: liquid
Color: Dark blue
Resistivity: 300 to 1000 ohm-cm
Viscosity: 5 - 11 cP
pH: 5- 11
Surface Tension: 33-40 dyne/cm
Work Function: -4.8 eV (Scanning Kelvin Probe)
Density: ~0.95 g/cm3



Component 4: Plexcore® OC RG-1155 organic conductive ink
Form: liquid
Color: Dark blue
Resistivity: 300 to 1000 ohm-cm
Viscosity: 5 - 11 cP
pH: 5 - 11
Surface Tension: 33-40 dyne/cm
Work Function: -5.25 eV (Scanning Kelvin Probe)
Density: ~0.95 g/cm3
Printed electronics (PE) exploits the ability of organic materials to be solution processed onto large area substrates at a fraction of the manufacturing cost associated with conventional vapor deposition techniques. One of the unique features of Plexcore OC inks is the ability to be tuned to match the work function, resistivity, and viscosity, for example, required for specific device architectures and application techniques. This tunability provides the versatility needed to develop new solid state lighting (SSL) device stacks. Plextronics′ Plexcore® technology platform is comprised of semiconducting polymers and solution-processable inks that are designed to maximize the efficiency, lifetime and stability of printed electronics (PE) devices.[1]

Application

Organic Conductive Inks for printed electronics applications including OLED devices.

Legal Information

Plexcore is a registered trademark of Plextronics, Inc.

Kit Components Also Available Separately

Product No.
Description
SDS

  • 719110Plexcore® OC RG-1110 organic conductive ink 10 mLSDS

  • 719129Plexcore® OC RG-1115 organic conductive ink 10 mLSDS

  • 719137Plexcore® OC RG-1150 organic conductive ink 10 mLSDS

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymer-based Materials for Printed Electronics: Enabling High Efficiency Solar Power and Lighting
Tipnis R,et al.
Material Matters, 3(4), 92-92 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service