Skip to Content
Merck
All Photos(1)

Documents

718734

Sigma-Aldrich

5-Methoxy-2-nitro-4-(trifluoromethyl)phenylacetonitrile

97%

Synonym(s):

5-Methoxy-2-nitro-4-(trifluoromethyl)benzyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7F3N2O3
CAS Number:
Molecular Weight:
260.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

102-106 °C

SMILES string

COc1cc(CC#N)c(cc1C(F)(F)F)[N+]([O-])=O

InChI

1S/C10H7F3N2O3/c1-18-9-4-6(2-3-14)8(15(16)17)5-7(9)10(11,12)13/h4-5H,2H2,1H3

InChI key

IANAREFPKLOIAF-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service