Skip to Content
Merck
All Photos(1)

Key Documents

716375

Sigma-Aldrich

2,5-Dibromo-3-methylthiophene

97%

Synonym(s):

3-Methyl-2,5-dibromothiophene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€69.40

€69.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€69.40

About This Item

Empirical Formula (Hill Notation):
C5H4Br2S
CAS Number:
Molecular Weight:
255.96
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€69.40


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.613

density

1.974 g/mL at 25 °C

functional group

bromo

storage temp.

2-8°C

SMILES string

Cc1cc(Br)sc1Br

InChI

1S/C5H4Br2S/c1-3-2-4(6)8-5(3)7/h2H,1H3

InChI key

IHFXZROPBCBLLG-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service