Skip to Content
Merck
All Photos(1)

Key Documents

662232

Sigma-Aldrich

N-Heterocyclic Carbene Ligands Kit I

Synonym(s):

Heterocyclic Carbene Ligands

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€385.00

€385.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
€385.00

About This Item

UNSPSC Code:
12350000
NACRES:
NA.22

€385.00


Check Cart for Availability

Request a Bulk Order

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

Kit Components Also Available Separately

Product No.
Description
SDS

  • 6566231,3-Bis-(2,6-diisopropylphenyl)imidazolinium chloride, 97% 1 gSDS

  • 6566311,3-Bis(2,4,6-trimethylphenyl)imidazolinium chloride, 95% 1 gSDS

  • 6600191,3-Diisopropylimidazolium tetrafluoroborate, 96% 1 gSDS

  • 6600351,3-Bis(1-adamantyl)imidazolium tetrafluoroborate, 97% 1 gSDS

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

In collaboration with Umicore AG and Co.,1 we are pleased to offer a series of robust Pd(II) and Pd(0) complexes employed as the linchpin in C–C bond forming reactions.

Metal complex-catalyzed cross-coupling reactions of unactivated substrates introduce diverse phosphine ligands in chemical marketplace.

A wide range of NHC ligands are commonly available which exhibit high activities.

Questions

  1. What specific compounds are included in the N-Heterocyclic Carbene Ligands Kit I (662232)?

    1 answer
    1. The kit components can be found below the 'DESCRIPTION' section under 'Kit Components Also Available Separately'. This kit contains the following:

      1,3-Bis-(2,6-diisopropylphenyl)imidazolinium chloride, 97% 1 g
      1,3-Bis(2,4,6-trimethylphenyl)imidazolinium chloride, 95% 1 g
      1,3-Diisopropylimidazolium tetrafluoroborate, 96% 1 g
      1,3-Bis(1-adamantyl)imidazolium tetrafluoroborate, 97% 1 g

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service