Skip to Content
Merck
All Photos(2)

Key Documents

715298

Sigma-Aldrich

1,2,4,5-Tetrakis(4-carboxyphenyl)benzene

greener alternative

contains up to 6 wt. % water, ≥98%

Synonym(s):

4,4′,4′′,4′′′-benzene-1,2,4,5-tetrayltetrabenzoic acid, H4TCPB

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€247.00

€247.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€247.00

About This Item

Empirical Formula (Hill Notation):
C34H22O8
Molecular Weight:
558.53
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

€247.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98%

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

SMILES string

OC(=O)c1ccc(cc1)-c2cc(-c3ccc(cc3)C(O)=O)c(cc2-c4ccc(cc4)C(O)=O)-c5ccc(cc5)C(O)=O

InChI

1S/C34H22O8/c35-31(36)23-9-1-19(2-10-23)27-17-29(21-5-13-25(14-6-21)33(39)40)30(22-7-15-26(16-8-22)34(41)42)18-28(27)20-3-11-24(12-4-20)32(37)38/h1-18H,(H,35,36)(H,37,38)(H,39,40)(H,41,42)

InChI key

SRTQKANXPMBQCX-UHFFFAOYSA-N

General description

1,2,4,5-Tetrakis(4-carboxyphenyl)benzene (H4TCPB) is an organic linker with a molecular structure that has four carboxyl phenyl groups that are symmetrically attached to the central benzene core. It can be synthesized by using a Suzuki coupling reaction.[1]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

(H4TCPB) can be used for a variety of applications such as:
  • the formation of supramolecular heterostructures to functionalize black phosphorus[1]
  • synthesis of microporous metal-organic frameworks (MOFs) for potential usage in hydrogen storage, gas separation, and catalysis.[2][3]

[1,1′:4′,1]Terphenyl- 3,3″,5,5″-tetracarboxylic acid (H4TPTC) is used as a linker molecule for the synthesis of high performance metal-organic frameworks (MOFs).[4]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and structure of Zr (iv)-and Ce (iv)-based CAU-24 with 1, 2, 4, 5-tetrakis (4-carboxyphenyl) benzene
Lammert M, et al.
Dalton Transactions, 45(47), 18822-18826 (2016)
Near-critical water, a cleaner solvent for the synthesis of a metal-organic framework
Ibarra IA, et al.
Green Chemistry, 14(1), 117-122 (2012)
Lilia S Xie et al.
Chemical reviews, 120(16), 8536-8580 (2020-04-11)
Metal-organic frameworks (MOFs) are intrinsically porous extended solids formed by coordination bonding between organic ligands and metal ions or clusters. High electrical conductivity is rare in MOFs, yet it allows for diverse applications in electrocatalysis, charge storage, and chemiresistive sensing
Singlet oxygen-mediated selective C--H bond hydroperoxidation of ethereal hydrocarbons
Sagadevan A, et al.
Nature Communications, 8(1), 1-8 (2017)
Selected Applications of Metal-Organic Frameworks in Sustainable Energy Technologies?.
Caskey SR and Matzger AJ.
Material Matters, 4(4), 111-111 (2009)

Articles

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service