Skip to Content
Merck
All Photos(2)

Key Documents

1.13861

Sigma-Aldrich

Dihexyl sodium sulfosuccinate

for synthesis

Synonym(s):

Dihexyl sodium sulfosuccinate, 2-Sulfosuccinic acid di-N-hexyl ester sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€679.00

€679.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
€679.00

About This Item

Empirical Formula (Hill Notation):
C16H29NaO7S
CAS Number:
Molecular Weight:
388.45
MDL number:
UNSPSC Code:
12352107
EC Index Number:
221-109-1
NACRES:
NA.22

€679.00


Estimated to ship on24 May 2025


Request a Bulk Order

Quality Level

form

solid

potency

1750 mg/kg LD50, oral (Rat)

pH

5-7 (25 °C, 50 g/L in H2O)

mp

230 °C (decomposes)

solubility

364 g/L

bulk density

180 kg/m3

storage temp.

15-25°C

SMILES string

O=S(C(C(OCCCCCC)=O)CC(OCCCCCC)=O)(O[Na])=O

InChI

1S/C16H30O7S.Na/c1-3-5-7-9-11-22-15(17)13-14(24(19,20)21)16(18)23-12-10-8-6-4-2;/h14H,3-13H2,1-2H3,(H,19,20,21);/q;+1/p-1

InChI key

WVFDILODTFJAPA-UHFFFAOYSA-M

Analysis Note

Assay (two-phase titration, cal. on anhydrous substance): ≥ 98.5 %
Identity (IR-spectrum): passes test
pH-value (5 %; water): 5 - 7
Water (according to Karl Fischer): 4 - 5

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service