Skip to Content
Merck
All Photos(1)

Key Documents

707767

Sigma-Aldrich

Uridine-15N2 5′-triphosphate disodium salt solution

100 mM (in 5mM Tris HCl / H2O), ≥98 atom % 15N, ≥95% (CP)

Synonym(s):

UTP-15N2 disodium salt solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€93.70
10 MG
€467.00
25 MG
€1,010.00

€93.70


Estimated to ship on13 April 2025


Request more information

Select a Size

Change View
1 MG
€93.70
10 MG
€467.00
25 MG
€1,010.00

About This Item

Empirical Formula (Hill Notation):
C9H1315N2Na2O15P3
Molecular Weight:
530.09
UNSPSC Code:
41106305
NACRES:
NA.12

€93.70


Estimated to ship on13 April 2025


Request more information

isotopic purity

≥98 atom % 15N

Quality Level

Assay

≥95% (CP)

form

solution

concentration

100 mM (in 5mM Tris HCl / H2O)

technique(s)

bio NMR: suitable

mass shift

M+2

storage temp.

−20°C

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Other Notes

Molecular weight and empirical formula based on the free acid.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service