Skip to Content
Merck
All Photos(2)

Key Documents

200921

Sigma-Aldrich

Silver(II) fluoride

≥98%

Synonym(s):

Silver difluoride

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€173.00

€173.00


Estimated to ship on03 April 2025


Request a Bulk Order

Select a Size

Change View
10 G
€173.00

About This Item

Empirical Formula (Hill Notation):
AgF2
CAS Number:
Molecular Weight:
145.87
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥98%
form:
powder

€173.00


Estimated to ship on03 April 2025


Request a Bulk Order

Quality Level

Assay

≥98%

form

powder

mp

690 °C (lit.)

density

4.57 g/mL at 25 °C (lit.)

SMILES string

F[Ag]F

InChI

1S/Ag.2FH/h;2*1H/q+2;;/p-2

InChI key

KWVVTSALYXIJSS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Silver (II) fluoride may be used for selective fluorination of carbon-hydrogen bond in pyridines and diazines. [1]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Selective CH fluorination of pyridines and diazines inspired by a classic amination reaction.
Fier PS & Hartwig JF.
Science, 342.6161, 956-960 (2013)

Articles

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service