701637
Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C16H24BF4Ir
CAS Number:
Molecular Weight:
495.38
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
description
store and handling under Argon gas at all time
reaction suitability
core: iridium
reagent type: catalyst
mp
32-48 °C
storage temp.
−20°C
SMILES string
[Ir+].F[B-](F)(F)F.C1CC=CCCC=C1.C2CC=CCCC=C2
InChI
1S/2C8H12.BF4.Ir/c2*1-2-4-6-8-7-5-3-1;2-1(3,4)5;/h2*1-2,7-8H,3-6H2;;/q;;-1;+1/b2*2-1-,8-7-;;
InChI key
NKIQXDAGMKBVPE-QMDOQEJBSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service