Skip to Content
Merck
All Photos(1)

Key Documents

299391

Sigma-Aldrich

Tetrairidium dodecacarbonyl

98%

Synonym(s):

Iridium carbonyl

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€214.00

€214.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€214.00

About This Item

Linear Formula:
Ir4(CO)12
CAS Number:
Molecular Weight:
1104.99
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder
solubility:
THF: slightly soluble(lit.)
toluene: slightly soluble(lit.)

€214.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

powder

reaction suitability

reagent type: catalyst
core: iridium

mp

195 °C (dec.) (lit.)

solubility

THF: slightly soluble(lit.)
toluene: slightly soluble(lit.)

SMILES string

[Ir].[Ir].[Ir].[Ir].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]

InChI

1S/12CO.4Ir/c12*1-2;;;;

InChI key

XWDKRVSSHIJNJP-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service