Skip to Content
Merck
All Photos(1)

Key Documents

697222

Sigma-Aldrich

Tantalum(V) oxide

−60 mesh, 99.5%

Synonym(s):

Tantalum pentoxide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€146.00

€146.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€146.00

About This Item

Linear Formula:
Ta2O5
CAS Number:
Molecular Weight:
441.89
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

€146.00


Check Cart for Availability

Request a Bulk Order

Assay

99.5%

form

powder

particle size

−60 mesh

density

8.2 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=[Ta](=O)O[Ta](=O)=O

InChI

1S/5O.2Ta

InChI key

PBCFLUZVCVVTBY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service