Skip to Content
Merck
All Photos(2)

Key Documents

204536

Sigma-Aldrich

Tantalum(V) oxide

<20 μm, 99.99% trace metals basis

Synonym(s):

Tantalum pentoxide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€111.00
50 G
€354.00

€111.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€111.00
50 G
€354.00

About This Item

Linear Formula:
Ta2O5
CAS Number:
Molecular Weight:
441.89
EC Number:
MDL number:
UNSPSC Code:
12352303
eCl@ss:
38180602
PubChem Substance ID:
NACRES:
NA.23

€111.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: tantalum

particle size

<20 μm

density

8.2 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=[Ta](=O)O[Ta](=O)=O

InChI

1S/5O.2Ta

InChI key

PBCFLUZVCVVTBY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ta2O5 (tantalum pentoxide) is a transition-metal oxide with a high refractive index (n=2.02–2.16), high dielectric constant, wide bandgap (4.0–4.5 eV), and wide transmission spectrum. It is a non-silicon dielectric material used in microelectronic devices and the ideal capacitive material for next-generation multi-chip modules. Due to its high acidity, it can serve as a catalyst for the oxidation of alkanes and pollution reduction. In addition, it is successfully used for the anode oxidization of electrodes in lithium batteries.

Application


  • Double-layered nanoparticle stacks for spectro-electrochemical applications.: This study discusses the development of double-layered nanoparticle stacks incorporating Tantalum(V) oxide, which have significant potential in spectro-electrochemical applications due to their high refractive index and dielectric properties (Frank et al., 2012).

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service