Skip to Content
Merck
All Photos(1)

Key Documents

695173

Sigma-Aldrich

Dichloro(2-pyridinecarboxylato)gold

Synonym(s):

(SP-4-3)-Dichloro(2-pyridinecarboxylato-κN1,κO2)gold

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€352.00

€352.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€352.00

About This Item

Empirical Formula (Hill Notation):
C6H4NAuCl2O2
CAS Number:
Molecular Weight:
389.97
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€352.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

reaction suitability

core: gold
reagent type: catalyst

mp

175-176 °C

SMILES string

Cl[Au]1(Cl)OC(=O)C2=[N]1C=CC=C2

InChI

1S/C6H5NO2.Au.2ClH/c8-6(9)5-3-1-2-4-7-5;;;/h1-4H,(H,8,9);;2*1H/q;+3;;/p-3

InChI key

PKAWSKSRAJXTIH-UHFFFAOYSA-K

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Access gold precatalysts, silver salts, and unsaturated building blocks to accelerate research success in catalysis applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service