Skip to Content
Merck
All Photos(1)

Key Documents

699616

Sigma-Aldrich

Chloro[tris(2,4-di-tert-butylphenyl)phosphite]gold

Synonym(s):

[Tris(2,4-di-tert-butylphenyl)phosphite]gold chloride

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€66.90

€66.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€66.90

About This Item

Empirical Formula (Hill Notation):
C42H63AuClO3P
CAS Number:
Molecular Weight:
879.34
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€66.90


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

reaction suitability

core: gold
reagent type: catalyst

mp

200-202 °C

SMILES string

Cl[Au].CC(C)(C)c1ccc(OP(Oc2ccc(cc2C(C)(C)C)C(C)(C)C)Oc3ccc(cc3C(C)(C)C)C(C)(C)C)c(c1)C(C)(C)C

InChI

1S/C42H63O3P.Au.ClH/c1-37(2,3)28-19-22-34(31(25-28)40(10,11)12)43-46(44-35-23-20-29(38(4,5)6)26-32(35)41(13,14)15)45-36-24-21-30(39(7,8)9)27-33(36)42(16,17)18;;/h19-27H,1-18H3;;1H/q;+1;/p-1

InChI key

GCWCLHMEYZKZRO-UHFFFAOYSA-M

Application

Gold Catalysts — 21st Century ′Gold Rush′

  • Intermolecular addition of carbon nucleophiles to 1,5 and 1,6 enynes[1]
Catalyst for:
  • [4C+2C] cycloadditions
  • Cis-selective single-cleavagte skeletal cycloisomerization
  • Diastereoselective Au-catalyzed intermolecular cyclopropanation
  • Elucidating the mechanism of "endocyclic" skeletal rearrangement of 1,6-enynes
  • Addition reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catelijne H M Amijs et al.
The Journal of organic chemistry, 73(19), 7721-7730 (2008-09-02)
Gold(I)-catalyzed addition of carbon nucleophiles to 1,6-enynes gives two different type of products by reaction at the cyclopropane or at the carbene carbons of the intermediate cyclopropyl gold carbenes. The 5-exo-dig cyclization is followed by most 1,6-enynes, although those bearing

Articles

Access gold precatalysts, silver salts, and unsaturated building blocks to accelerate research success in catalysis applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service