Skip to Content
Merck
All Photos(1)

Key Documents

689084

Sigma-Aldrich

2-(4-Benzyl-1-piperazinyl)-1-phenylethanone hydrobromide

96%

Synonym(s):

α-(4-Benzylpiperazinyl)acetophenone hydrobromide, N-Benzoylmethyl-N′-benzylpiperazine hydrobromide

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€135.00

€135.00


Estimated to ship on18 April 2025


Request a Bulk Order

Select a Size

Change View
500 MG
€135.00

About This Item

Empirical Formula (Hill Notation):
C19H22N2O · xHBr
CAS Number:
Molecular Weight:
294.39 (free base basis)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€135.00


Estimated to ship on18 April 2025


Request a Bulk Order

Quality Level

Assay

≥95.5% (HPLC)
96%

form

powder

impurities

≤5.0% water

functional group

ketone
phenyl

SMILES string

Br.O=C(CN1CCN(CC1)Cc2ccccc2)c3ccccc3

InChI

1S/C19H22N2O.BrH/c22-19(18-9-5-2-6-10-18)16-21-13-11-20(12-14-21)15-17-7-3-1-4-8-17;/h1-10H,11-16H2;1H

InChI key

BYGQOUUMIHHYAW-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service