Skip to Content
Merck
All Photos(1)

Documents

678848

Sigma-Aldrich

2-Aminomethyl-1,4-benzodioxane

97%

Synonym(s):

(2,3-Dihydrobenzo-1,4-dioxin-2-yl)methylamine, 1,4-Benzodioxane-2-methylamine, 2,3-Dihydro-1,4-benzodioxin-2-methanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11NO2
CAS Number:
Molecular Weight:
165.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.5594

bp

105-110 °C/3 mmHg

density

1.1679 g/mL at 25 °C

SMILES string

NCC1COc2ccccc2O1

InChI

1S/C9H11NO2/c10-5-7-6-11-8-3-1-2-4-9(8)12-7/h1-4,7H,5-6,10H2

InChI key

JHNURUNMNRSGRO-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service