Skip to Content
Merck
All Photos(1)

Key Documents

688770

Sigma-Aldrich

tert-Butyl acetoacetate

Arxada quality, ≥98 % (w/w) (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2COOC(CH3)3
CAS Number:
Molecular Weight:
158.19
Beilstein:
1680303
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

quality

Arxada quality

manufacturer/tradename

Arxada AG

concentration

≥98 % (w/w) (GC)

impurities

≤0.1% water
≤0.5% acid (as acetic acid)
≤2.0% tert-butanol

color

clear
APHA: ≤100

refractive index

n20/D 1.419 (lit.)

bp

71-72 °C/11 mmHg (lit.)

density

0.954 g/mL at 25 °C (lit.)

SMILES string

CC(=O)CC(=O)OC(C)(C)C

InChI

1S/C8H14O3/c1-6(9)5-7(10)11-8(2,3)4/h5H2,1-4H3

InChI key

JKUYRAMKJLMYLO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Xie et al.
Oncotarget, 5(20), 10048-10057 (2014-10-11)
Telomerase activation through the induction of its catalytic component TERT is essential in carcinogenesis. The regulatory mechanism and clinical significance underlying cancer-specific TERT expression have been extensively investigated in various human malignancies, but little is known about these in Merkel
Jordan M Spatz et al.
The Journal of biological chemistry, 290(27), 16744-16758 (2015-05-09)
Although bone responds to its mechanical environment, the cellular and molecular mechanisms underlying the response of the skeleton to mechanical unloading are not completely understood. Osteocytes are the most abundant but least understood cells in bones and are thought to
Ying Wang et al.
Endocrinology, 155(5), 1970-1981 (2014-03-08)
FSH is an essential regulator of mammalian reproduction. Its synthesis by pituitary gonadotrope cells is regulated by multiple endocrine and paracrine factors, including TGFβ superfamily ligands, such as the activins and inhibins. Activins stimulate FSH synthesis via transcriptional regulation of
Joseph G Crompton et al.
Cancer research, 75(2), 296-305 (2014-11-30)
Adoptive cell therapy (ACT) using autologous tumor-infiltrating lymphocytes (TIL) results in complete regression of advanced cancer in some patients, but the efficacy of this potentially curative therapy may be limited by poor persistence of TIL after adoptive transfer. Pharmacologic inhibition
Hiroshi Ono et al.
Cancer medicine, 3(3), 632-642 (2014-03-05)
Small-cell lung cancer (SCLC) is a subtype of lung cancer with poor prognosis. To identify accurate predictive biomarkers and effective therapeutic modalities, we focus on a long noncoding RNA, Hox transcript antisense intergenic RNA (HOTAIR), and investigated its expression, cellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service