Skip to Content
Merck
All Photos(1)

Key Documents

691380

Sigma-Aldrich

2,2,6-Trimethyl-4H-1,3-dioxin-4-one

Arxada quality, ≥93.0% (GC)

Synonym(s):

TMD, Diketene acetone adduct

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€462.00
25 KG
€3,480.00
180 KG
€16,540.00

€462.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
€462.00
25 KG
€3,480.00
180 KG
€16,540.00

About This Item

Empirical Formula (Hill Notation):
C7H10O3
CAS Number:
Molecular Weight:
142.15
Beilstein:
2408
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€462.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥93.0% (GC)

form

liquid

quality

Arxada quality

manufacturer/tradename

Arxada AG

impurities

≤0.10% diketene (GC)
≤0.4% dehydracetic acid (GC)
≤0.5% 4-methyl-3-penten-2-one (GC)
≤6.0% acetone (GC)

refractive index

n20/D 1.460 (lit.)

bp

~275 °C (lit.)
65-67 °C/2 mmHg (lit.)

mp

12-13 °C (lit.)

solubility

H2O: insoluble

density

1.07 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1=CC(=O)OC(C)(C)O1

InChI

1S/C7H10O3/c1-5-4-6(8)10-7(2,3)9-5/h4H,1-3H3

InChI key

XFRBXZCBOYNMJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,2,6-Trimethyl-4H-1,3-dioxin-4-one is generally used as a reactant for the synthesis of acetoacetic esters from aliphatic as well as aromatic alcohols.[1][2] It can also acetoacetylate amines and thiols.[3]

Caution

products darkens with time

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and biological evaluation of some new 1, 4-dihydropyridines containing different ester substitute and diethyl carbamoyl group as anti-tubercular agents
Khoshneviszadeh M, et al.
Bioorganic & Medicinal Chemistry, 17(4), 1579-1586 (2009)
Synthesis, study of 3D structures, and pharmacological activities of lipophilic nitroimidazolyl-1, 4-dihydropyridines as calcium channel antagonist
Miri R, et al.
Bioorganic & Medicinal Chemistry, 14(14), 4842-4849 (2006)
Acetoacetylation with 2, 2, 6-trimethyl-4H-1, 3-dioxin-4-one: a convenient alternative to diketene.
Clemens RJ and Hyatt JA.
The Journal of Organic Chemistry, 50(14), 2431-2435 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service