Skip to Content
Merck
All Photos(2)

Key Documents

686247

Sigma-Aldrich

Zirconium carboxyethyl acrylate

60% (n-propanol)

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€657.00

€657.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€657.00

About This Item

Linear Formula:
Zr(CH2CHCO2CH2CH2CO2)4
CAS Number:
Molecular Weight:
663.69
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€657.00


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

contains

500 ppm methyl hydroquinone as inhibitor

reaction suitability

reagent type: catalyst
core: zirconium

concentration

60% (n-propanol)

bp

100-121 °C

density

1.101 g/mL at 25 °C

SMILES string

C=CC(=O)OCCC(=O)O[Zr](OC(=O)CCOC(=O)C=C)(OC(=O)CCOC(=O)C=C)OC(=O)CCOC(=O)C=C

InChI

1S/4C6H8O4.Zr/c4*1-2-6(9)10-4-3-5(7)8;/h4*2H,1,3-4H2,(H,7,8);/q;;;;+4/p-4

InChI key

XQEXAYLRUODZQC-UHFFFAOYSA-J

Application

Please see product no. 686204 for additional application and usage information (Aldrich Technical Bulletin AL-238)
Zirconium-containing multifunctional acrylate useful for producing cured, transparent films with high refractive indices.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

59.0 °F - closed cup - Solvent

Flash Point(C)

15 °C - closed cup - Solvent

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service