Skip to Content
Merck
All Photos(1)

Key Documents

592439

Sigma-Aldrich

Pentabromophenyl methacrylate

96%

Synonym(s):

2,3,4,5,6-Pentafluorostyrene polymer, Poly(2,3,4,5,6-pentafluorostyrene)

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€347.00

€347.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€347.00

About This Item

Empirical Formula (Hill Notation):
C10H5Br5O2
CAS Number:
Molecular Weight:
556.67
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€347.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

mp

139-141 °C (lit.)

storage temp.

−20°C

SMILES string

CC(=C)C(=O)Oc1c(Br)c(Br)c(Br)c(Br)c1Br

InChI

1S/C10H5Br5O2/c1-3(2)10(16)17-9-7(14)5(12)4(11)6(13)8(9)15/h1H2,2H3

InChI key

OFZRSOGEOFHZKS-UHFFFAOYSA-N

Application

Monomer used to make high refractive index polymer for optical waveguides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olesya Daikos et al.
Physical chemistry chemical physics : PCCP, 18(47), 32369-32377 (2016-11-18)
Pentabrominated and fluorinated aromatic (meth)acrylates as well as their non-halogenated counterparts have been studied with the aim to avoid conventional photoinitiators and to overcome some negative consequences related to their use. Therefore, RTIR spectroscopy, laser flash photolysis and GC/MS were

Articles

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service