Skip to Content
Merck
All Photos(1)

Key Documents

679909

Sigma-Aldrich

Potassium

cubes (in mineral oil), 99.5% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
K
CAS Number:
Molecular Weight:
39.10
MDL number:
UNSPSC Code:
12141804
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

0.09 mmHg ( 260 °C)

Assay

99.5% trace metals basis

form

cubes (in mineral oil)

reaction suitability

reagent type: reductant

resistivity

6.1 μΩ-cm, 20°C

bp

760 °C (lit.)

mp

64 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

SMILES string

[K]

InChI

1S/K

InChI key

ZLMJMSJWJFRBEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Combines under pressure with silver metal to form 2 new compounds: K2Ag and K3Ag.†

Quantity

1 cube = ~20g

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole Schmitt et al.
Physiological reviews, 94(2), 609-653 (2014-04-03)
About 10 distinct potassium channels in the heart are involved in shaping the action potential. Some of the K+ channels are primarily responsible for early repolarization, whereas others drive late repolarization and still others are open throughout the cardiac cycle.
Glucose-insulin-potassium revived: current status in acute coronary syndromes and the energy-depleted heart.
Alexandra N Grossman et al.
Circulation, 127(9), 1040-1048 (2013-03-06)
Jordan E Bisanz et al.
Applied and environmental microbiology, 81(15), 4965-4975 (2015-05-17)
The nutritional status of pregnant women is vital for healthy outcomes and is a concern for a large proportion of the world's population. The role of the microbiota in pregnancy and nutrition is a promising new area of study with
Stefan D Anker et al.
European journal of heart failure, 17(10), 1050-1056 (2015-05-27)
Hyperkalaemia in heart failure patients limits use of cardioprotective renin-angiotensin-aldosterone system inhibitors (RAASi). Sodium zirconium cyclosilicate (ZS-9) is a selective potassium ion trap, whose mechanism of action may allow for potassium binding in the upper gastrointestinal tract as early as
Mayumi Tada et al.
The European journal of neuroscience, 39(11), 1720-1728 (2014-01-11)
Calcium imaging of individual neurons is widely used for monitoring their activity in vitro and in vivo. Synthetic fluorescent calcium indicator dyes are commonly used, but the resulting calcium signals sometimes suffer from a low signal-to-noise ratio (SNR). Therefore, it

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service