Skip to Content
Merck
All Photos(1)

Key Documents

3860-OP

Millipore

DTT

OmniPur® Grade, >= 99.4%, Used for reduction of protein disulfide linkages

Synonym(s):

OmniPur® DTT, Cleland′s Reagent, Dithiothreitol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
Inquire
25 G
Inquire


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
5 G
Inquire
25 G
Inquire

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.28


Estimated to ship on01 June 2025


Request a Bulk Order

grade

Molecular Biology

Quality Level

Assay

≥99.4%

form

crystalline powder

reaction suitability

reagent type: reductant

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 5%

foreign activity

DNAse, none detected
Protease, none detected
RNAse, none detected

shipped in

wet ice

storage temp.

−20°C

SMILES string

SC[C@H](O)[C@@H](O)CS

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/t3-,4-/m0/s1

InChI key

VHJLVAABSRFDPM-IMJSIDKUSA-N

General description

Used for reduction of protein disulfide linkages. Molecular Biology

A (0.02 M in water): ≤0.05 AU
Assay (-SH content): ≥99.4%
DNAse: none detected
Infrared spectrum: Conforms to standard
Loss on drying: ≤0.5%
Melting point: 39-43°C
Protease: none detected
RNAse: none detected
Solubility (5%, water): To pass test





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application


  • Oxidative potential of particulate matter and its association to respiratory health endpoints in high-altitude cities in Bolivia.: This research investigates the oxidative potential of particulate matter and its impact on respiratory health in Bolivian high-altitude cities. DTT (Dithiothreitol) is used as a reducing agent to measure the oxidative potential of particulates, providing insights into their health impacts and guiding environmental policy (Borlaza-Lacoste et al., 2024).

  • Protein purification via consecutive histidine-polyphosphate interaction.: This study explores an innovative method for protein purification using DTT to maintain reducing conditions essential for the preservation of protein structures. The findings highlight DTT′s role in enhancing the efficiency and yield of protein purification processes, which is critical for biochemical research and industrial applications (Zhou et al., 2024).

Warning

Toxicity: Harmful (C)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service