Skip to Content
Merck
All Photos(2)

Key Documents

679798

Sigma-Aldrich

Bis(ethylcyclopentadienyl)ruthenium(II)

packaged for use in deposition systems

Synonym(s):

Ru(EtCp)2, Diethylruthenocene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C7H9RuC7H9
CAS Number:
Molecular Weight:
287.36
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

liquid

reaction suitability

core: ruthenium
reagent type: catalyst

refractive index

n20/D 1.5870 (lit.)

bp

100 °C/0.01 mmHg (lit.)

mp

6 °C (lit.)

density

1.3412 g/mL at 25 °C (lit.)

SMILES string

[Ru].CC[C]1[CH][CH][CH][CH]1.CC[C]2[CH][CH][CH][CH]2

InChI

1S/2C7H9.Ru/c2*1-2-7-5-3-4-6-7;/h2*3-6H,2H2,1H3;

InChI key

VLTZUJBHIUUHIK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 44 Ruthenium

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service