Skip to Content
Merck
All Photos(1)

Documents

510645

Sigma-Aldrich

Bis(ethylcyclopentadienyl)cobalt(II)

Synonym(s):

(EtCp)2Co, 1,1′-Diethylcobaltocene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18Co
CAS Number:
Molecular Weight:
245.23
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

liquid

reaction suitability

core: cobalt

density

1.516 g/mL at 25 °C (lit.)

SMILES string

[Co].CC[C]1[CH][CH][CH][CH]1.CC[C]2[CH][CH][CH][CH]2

InChI

1S/2C7H9.Co/c2*1-2-7-5-3-4-6-7;/h2*3-6H,2H2,1H3;

InChI key

YHCQFTZSIGZRTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 27 Cobalt

Application

This metallocene compound may be encapsulated in single walled carbon nanotubes, resulting in a tailored carbon nano structure of specific electronic functionality.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Diameter-selective encapsulation of metallocenes in single-walled carbon nanotubes.
Li L-J, et al.
Nature Materials, 4.6, 481-485 (2005)

Articles

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

High Purity Metalorganic Precursors for CPV Device Fabrication

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service