Skip to Content
Merck
All Photos(1)

Key Documents

679232

Sigma-Aldrich

Trimethoxymethylsilane

deposition grade, ≥98%

Synonym(s):

Methyltrimethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3Si(OCH3)3
CAS Number:
Molecular Weight:
136.22
Beilstein:
1736151
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

deposition grade

Assay

≥98%

form

liquid

refractive index

n20/D 1.371 (lit.)

bp

102-104 °C (lit.)

density

0.955 g/mL at 25 °C (lit.)

SMILES string

CO[Si](C)(OC)OC

InChI

1S/C4H12O3Si/c1-5-8(4,6-2)7-3/h1-4H3

InChI key

BFXIKLCIZHOAAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

48.2 °F

Flash Point(C)

9 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Yan Zhang et al.
Guang pu xue yu guang pu fen xi = Guang pu, 26(9), 1631-1635 (2006-11-23)
The characters of optical sensing films for ammonia were investigated based on a fluorescent indicator aminofluorescein (AF) immobilization. An organically modified silicates (ORMOSILSs) obtained by copolymerizing various proportions of methyltrimethoxysilane (Me-TriMOS) and tetramethoxysilane (TMOS) was applied. Compared with the fluorescent
Yan Gao et al.
Langmuir : the ACS journal of surfaces and colloids, 26(2), 1239-1246 (2009-09-03)
The immobilization of biological species such as proteins and enzymes in sol-gel hosts is currently an area of intense research activity. However, the majority of these studies have been directed toward investigating the biological activity or physicochemical properties of the
M Morpurgo et al.
Farmaco (Societa chimica italiana : 1989), 60(8), 675-683 (2005-06-28)
The influence of processing parameters and synthetic strategies in the properties of sol-gel derived silica matrices intended for the release of bioactive compounds was investigated. The time-evolution of the matrix properties during its aging at room temperature in the dry
Mary E Robbins et al.
Journal of the American Chemical Society, 125(20), 6068-6069 (2003-06-06)
The release of nitric oxide (NO) from polymers has proven to be highly effective at inhibiting platelet adhesion and thus enhancing the blood compatibility of medical implants. Micropatterning techniques were used to design surfaces that release NO while preserving the
Bao Chau N Nguyen et al.
ACS applied materials & interfaces, 2(5), 1430-1443 (2010-04-30)
The elastic properties and/or flexibility of polymer reinforced silica aerogels having methyltrimethoxysilane (MTMS) and bis(trimethoxysilylpropyl)amine (BTMSPA) making up the silica structure are examined. The dipropylamine spacer from BTMSPA is used both to provide a flexible linking group in the silica

Articles

Deposition Grade Silanes, fully characterized by chemical analysis and nuclear magnetic resonance (NMR) with greater than 98% purity, for Sol-Gel Processes.

Reactive silicone chemistry: Focus on pure silicon production, polymerizations, and controlled stereochemistry reactions.

atomic layer deposition (ALD), microelectronics, Mo:Al2O3 films, nanocomposite coating, photovoltaics, semiconductor devices, W:Al2O3 films, composite films, layer-by-layer

Silica's versatility spans various industries, including biomedical applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service