Skip to Content
Merck
All Photos(1)

Documents

675261

Sigma-Aldrich

2-Hydroxyisobutyric acid-13C4

99 atom % 13C, 98% (CP)

Synonym(s):

2-Hydroxy-2-(methyl-13C)-propanoic-1,2,3-13C3 acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH313C(OH)13CH313COOH
CAS Number:
Molecular Weight:
108.08
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mp

76-80 °C

mass shift

M+4

SMILES string

[13CH3][13C]([13CH3])(O)[13C](O)=O

InChI

1S/C4H8O3/c1-4(2,7)3(5)6/h7H,1-2H3,(H,5,6)/i1+1,2+1,3+1,4+1

InChI key

BWLBGMIXKSTLSX-JCDJMFQYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service