Skip to Content
Merck
All Photos(1)

Key Documents

164976

Sigma-Aldrich

α-Hydroxyisobutyric acid

98%

Synonym(s):

alpha-Hydroxyisobutyric acid, 2-Hydroxy-2-methylpropionic acid, 2-Methyllactic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€39.20

€39.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€39.20

About This Item

Linear Formula:
(CH3)2C(OH)COOH
CAS Number:
Molecular Weight:
104.10
Beilstein:
1744739
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€39.20


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

bp

84 °C/1.5 mmHg (lit.)

mp

76-80 °C (lit.)

solubility

water: soluble 10%, clear to very slightly hazy, colorless

functional group

carboxylic acid
hydroxyl

SMILES string

CC(C)(O)C(O)=O

InChI

1S/C4H8O3/c1-4(2,7)3(5)6/h7H,1-2H3,(H,5,6)

InChI key

BWLBGMIXKSTLSX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Hydroxyisobutyric acid (2-hydroxyisobutyric acid) is the building block for polymer synthesis[1]. It forms complexes with Eu(III) and has been studied by time resolved fluorescence spectroscopy[2].

Application

α-Hydroxyisobutyric acid was used as background electrolyte for separation and detection of the metal ions by on-line cyclic voltammetry[3].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Experimental factors in pulsed electrochemical detection in capillary electrophoresis.
Wen J, et al.
Journal of Chromatography A, 811(1), 181-192 (1998)
A Amberg et al.
Toxicological sciences : an official journal of the Society of Toxicology, 51(1), 1-8 (1999-09-25)
Methyl-tert-butyl ether (MTBE) is widely used as an additive to gasoline to increase oxygen content and reduce tail pipe emission of pollutants. Therefore, widespread human exposure may occur. To contribute to the characterization of potential adverse effects of MTBE, its
Thore Rohwerder et al.
Applied and environmental microbiology, 72(6), 4128-4135 (2006-06-06)
Fuel oxygenates such as methyl and ethyl tert-butyl ether (MTBE and ETBE, respectively) are degraded only by a limited number of bacterial strains. The aerobic pathway is generally thought to run via tert-butyl alcohol (TBA) and 2-hydroxyisobutyrate (2-HIBA), whereas further
Ana Carolina O Costa et al.
Journal of chromatography. A, 1171(1-2), 140-143 (2007-10-09)
The aim of this work was to develop a fast method using capillary electrophoresis for the determination of creatinine in human urine samples. The pH and constituents of the background electrolyte were selected by inspection of effective mobility of creatinine
Nadya Yaneva et al.
The Journal of biological chemistry, 287(19), 15502-15511 (2012-03-22)
Coenzyme B(12)-dependent acyl-CoA mutases are radical enzymes catalyzing reversible carbon skeleton rearrangements in carboxylic acids. Here, we describe 2-hydroxyisobutyryl-CoA mutase (HCM) found in the bacterium Aquincola tertiaricarbonis as a novel member of the mutase family. HCM specifically catalyzes the interconversion

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service