Skip to Content
Merck
All Photos(1)

Key Documents

671177

Sigma-Aldrich

Tributylmethylammonium methyl carbonate solution

~50% in methanol: water (2:3)

Synonym(s):

Methyltributylammonium methyl carbonate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€213.00

€213.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€213.00

About This Item

Empirical Formula (Hill Notation):
C15H33NO3
CAS Number:
Molecular Weight:
275.43
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€213.00


Check Cart for Availability

Request a Bulk Order

concentration

~50% in methanol: water (2:3)

functional group

amine

SMILES string

COC([O-])=O.CCCC[N+](C)(CCCC)CCCC

InChI

1S/C13H30N.C2H4O3/c1-5-8-11-14(4,12-9-6-2)13-10-7-3;1-5-2(3)4/h5-13H2,1-4H3;1H3,(H,3,4)/q+1;/p-1

InChI key

ZKWSMHMGQLTZGI-UHFFFAOYSA-M

Application

Tributylmethylammonium methyl carbonate can be used in the preparation of counter-cation anionic monomers by reacting with 2-acrylamido-2- methylpropanesulfonic acid and acrylic acid.[1]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.6 °F

Flash Point(C)

57 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular Design and Characterization of Ionic Monomers with Varying Ion Pair Interaction Energies
Matsuno R, et al.
Macromolecules, 53(5), 1629-1637 (2020)

Articles

CBILS© offers a simple, elegant synthetic method for the production of ionic liquids, developed by the Austrian company proionic.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service