Skip to Content
Merck
All Photos(1)

Key Documents

667811

Sigma-Aldrich

(−)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€111.00
500 MG
€385.00

€111.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€111.00
500 MG
€385.00

About This Item

Empirical Formula (Hill Notation):
C34H36P2
CAS Number:
Molecular Weight:
506.60
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€111.00


Check Cart for Availability

Request a Bulk Order

form

solid

mp

143-147 °C

functional group

phenyl
phosphine

SMILES string

C1C[C@@H](P(CCP2[C@H](CC[C@@H]2c3ccccc3)c4ccccc4)[C@H]1c5ccccc5)c6ccccc6

InChI

1S/C34H36P2/c1-5-13-27(14-6-1)31-21-22-32(28-15-7-2-8-16-28)35(31)25-26-36-33(29-17-9-3-10-18-29)23-24-34(36)30-19-11-4-12-20-30/h1-20,31-34H,21-26H2/t31-,32-,33-,34-/m1/s1

InChI key

VHHAZLMVLLIMHT-YFRBGRBWSA-N

Application

(−)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane can be used:
  • As a chiral ligand in the 1,3-dipolar cycloaddition reaction of azomethyne yielded [70]fullerene enantiomers; catalyzed by silver.[1]
  • As a component of the catalytic system along with silver, which is used for the synthesis of fullerene hybrids.[2]

Legal Information

Sold in collaboration with Namena Corp. for research purposes only.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hierarchical Selectivity in Fullerenes: Site-, Regio-, Diastereo-, and Enantiocontrol of the 1, 3-Dipolar Cycloaddition to C70
Maroto EE, et al.
Angewandte Chemie (International Edition in English), 50(27), 6060-6064 (2011)
Stereodivergent-at-Metal Synthesis of [60] Fullerene Hybrids
Marco-Martinez J, et al.
Angewandte Chemie (International Edition in English), 56(8), 2136-2139 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service