Skip to Content
Merck
All Photos(2)

Key Documents

670359

Sigma-Aldrich

Acetylthiomethyl-diphenylphosphine borane complex

≥98.0%

Synonym(s):

(T-4)-[S-[(Diphenylphosphino-κP)methyl] ethanethioate]trihydroboron

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18BOPS
CAS Number:
Molecular Weight:
288.15
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

form

solid

reaction suitability

reaction type: click chemistry
reagent type: ligand
reaction type: Staudinger Reaction

mp

52-55 °C

functional group

phosphine

storage temp.

2-8°C

SMILES string

B.CC(=O)SCP(c1ccccc1)c2ccccc2

InChI

1S/C15H15OPS.BH3/c1-13(16)18-12-17(14-8-4-2-5-9-14)15-10-6-3-7-11-15;/h2-11H,12H2,1H3;1H3

InChI key

MXPNVFCCEGQGEN-UHFFFAOYSA-N

Application

  • Traceless Staudinger ligation reagent with borane protecting group.
  • The borane group stabilizes the phosphine against oxidation and can be easily removed with mild basic or acidic conditions to yield the active phosphine.
  • After reaction with an azide, the phosphine is eliminated in the presence of water to yield a native amide bond.
  • Used in the synthesis of cyclic peptides.
Acetylthiomethyl-diphenylphosphine borane complex Ligation with DABCO

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Sold under license for research and development purposes only. US Patent 6,974,884 and related patents apply

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew B Soellner et al.
The Journal of organic chemistry, 67(14), 4993-4996 (2002-07-06)
The Staudinger ligation of peptides with a C-terminal phosphinothioester and N-terminal azide is an emerging method in protein chemistry. Here, the first Staudinger ligations of nonglycyl azides are reported and shown to proceed both in nearly quantitative yield and with
Michaela Mühlberg et al.
Bioorganic & medicinal chemistry, 18(11), 3679-3686 (2010-05-15)
The traceless Staudinger ligation has recently found various applications in the field of peptide synthesis and modification, including immobilization and cyclization strategies. In this report, we utilize the traceless Staudinger ligation in the formation of amide bonds, which allows the

Related Content

Raines lab innovations include traceless Staudinger ligation and DTBA reagent, advancing chemical biology research with Sigma-Aldrich.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service