Skip to Content
Merck
All Photos(1)

Key Documents

667544

Sigma-Aldrich

Lithium chloride solution

0.5 M in anhydrous tetrahydrofuran

Synonym(s):

Lithium chloride, Lithium monochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiCl
CAS Number:
Molecular Weight:
42.39
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

concentration

0.5 M in anhydrous tetrahydrofuran

impurities

≤50 ppm water

density

0.913 g/mL at 25 °C

SMILES string

[Li+].[Cl-]

InChI

1S/ClH.Li/h1H;/q;+1/p-1

InChI key

KWGKDLIKAYFUFQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Lithium chloride (LiCl) is a source of nucleophilic chloride anion and it can convert alcohols to alkyl chlorides.[1] It catalyzes the ortholithiations of a range of arenes mediated by lithium diisopropylamide (LDA).[2] LiCl solution is also used in the preparation of organozinc halides.[3]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lithium Chloride.
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)
On-demand synthesis of organozinc halides under continuous flow conditions.
Berton, Mateo et al.
Nature Protocols, 13(1), 324-324 (2018)
Lekha Gupta et al.
The Journal of organic chemistry, 74(5), 2231-2233 (2009-02-05)
Ortholithiations of a range of arenes mediated by lithium diisopropylamide (LDA) in THF at -78 degrees C reveal substantial accelerations by as little as 0.5 mol % of LiCl (relative to LDA). Substrate dependencies suggest a specific range of reactivity
Qi Wu et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(36), 14765-14770 (2013-08-22)
Diphtheria toxin-mediated, acute ablation of hypothalamic neurons expressing agouti-related protein (AgRP) in adult mice leads to anorexia and starvation within 7 d that is caused by hyperactivity of neurons within the parabrachial nucleus (PBN). Because NMDA glutamate receptors are involved
Karla M Acevedo et al.
The Journal of biological chemistry, 289(16), 11007-11019 (2014-03-13)
Amyloid precursor protein (APP) undergoes post-translational modification, including O- and N-glycosylation, ubiquitination, and phosphorylation as it traffics through the secretory pathway. We have previously reported that copper promotes a change in the cellular localization of APP. We now report that

Articles

PEPPSI palladium N-heterocyclic-carbene catalyst system enhances efficiency and functional group tolerance in catalysis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service