Merck
  • Lithium diisopropylamide-mediated ortholithiations: lithium chloride catalysis.
Lithium diisopropylamide-mediated ortholithiations: lithium chloride catalysis.
The Journal of organic chemistry (2009-02-05)
Lekha Gupta, Alexander C Hoepker, Kanwal J Singh, David B Collum
ABSTRACT
Ortholithiations of a range of arenes mediated by lithium diisopropylamide (LDA) in THF at -78 degrees C reveal substantial accelerations by as little as 0.5 mol % of LiCl (relative to LDA). Substrate dependencies suggest a specific range of reactivity within which the LiCl catalysis is optimal. Standard protocols with unpurified commercial samples of n-butyllithium to prepare LDA or commercially available LDA show marked batch-dependent rates--up to 100-fold--that could prove significant to the unwary practitioner. Other lithium salts elicit more modest accelerations. The mechanism is not discussed.
Product Description
Supelco
Diisopropylamine, analytical standard
Sigma-Aldrich
Diisopropylamine, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Lithium diisopropylamide, 97%
Sigma-Aldrich
Diisopropylamine, purified by redistillation, 99.95%
Sigma-Aldrich
Lithium diisopropylamide solution, 2.0 M in THF/heptane/ethylbenzene
Sigma-Aldrich
Diisopropylamine, ≥99.5%
Sigma-Aldrich
n-Butyllithium solution, 2.5 M in hexanes
Sigma-Aldrich
n-Butyllithium solution, 11.0 M in hexanes