662518
3,4-Dimethoxythiophenol
97%
Synonym(s):
3,4-Dimethoxybenzenethiol
About This Item
Recommended Products
Assay
97%
refractive index
n20/D 1.591
density
1.175 g/mL at 25 °C
SMILES string
COc1ccc(S)cc1OC
InChI
1S/C8H10O2S/c1-9-7-4-3-6(11)5-8(7)10-2/h3-5,11H,1-2H3
InChI key
MTKAJLNGIVXZIS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service