Skip to Content
Merck
All Photos(1)

Key Documents

662275

Sigma-Aldrich

Trimethoxy(propyl)silane

97%

Synonym(s):

Propyltrimethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2Si(OCH3)3
CAS Number:
Molecular Weight:
164.27
Beilstein:
2070458
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.15 psi ( 20 °C)

Quality Level

Assay

97%

refractive index

n20/D 1.391 (lit.)

bp

142 °C (lit.)

density

0.932 g/mL at 25 °C (lit.)

SMILES string

CCC[Si](OC)(OC)OC

InChI

1S/C6H16O3Si/c1-5-6-10(7-2,8-3)9-4/h5-6H2,1-4H3

InChI key

HQYALQRYBUJWDH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huiying Ma et al.
Sensors (Basel, Switzerland), 19(24) (2019-12-15)
We proposed a coding and decoding method of suspension array (SA) based on micro-quartz pieces (MQPs) with different optical thicknesses. The capture probes (cDNA) were grafted onto the surfaces of MQPs and specifically recognized and combined with the partial sequence
Daniel K Macharia et al.
Journal of photochemistry and photobiology. B, Biology, 174, 10-17 (2017-07-28)
The simultaneous imaging and photothermal therapy of tumors have attracted much attention, and a prerequisite is to obtain multifunctional nanomaterials. Ideally, one kind of nanoparticles with single component can be used as both imaging agent and photothermal agent. Herein, we
Jing Sun et al.
Biotechnology and applied biochemistry, 65(3), 362-371 (2017-12-10)
In recent years, the application of carbonic anhydrase (CA) in CO2 removal has attracted great interest. However, obtaining high enzyme recovery activity is difficult in existing immobilization techniques. In this work, water plasma-treated poly(vinylidene fluoride) (PVDF) membranes were modified via
Paige J LeValley et al.
Colloids and surfaces. B, Biointerfaces, 174, 483-492 (2018-11-30)
Circulating tumor cells (CTCs) play a central role in cancer metastasis and represent a rich source of data for cancer prognostics and therapeutic guidance. Reliable CTC recovery from whole blood therefore promises a less invasive and more sensitive approach to
Yunyun Wang et al.
Journal of separation science, 41(8), 1880-1887 (2018-02-02)
An imprinted fluorescent sensor was fabricated based on SiO2 nanoparticles encapsulated with a molecularly imprinted polymer containing allyl fluorescein. High fluorine cypermethirin as template molecules, methyl methacrylate as functional monomer, and allyl fluorescein as optical materials synthesized a core-shell fluorescent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service