Skip to Content
Merck
All Photos(1)

Key Documents

64940

Sigma-Aldrich

N-Methoxycarbonylmaleimide

≥97.0% (N)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO4
CAS Number:
Molecular Weight:
155.11
Beilstein:
1450638
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97.0% (N)

form

solid

mp

63-67 °C

storage temp.

2-8°C

SMILES string

COC(=O)N1C(=O)C=CC1=O

InChI

1S/C6H5NO4/c1-11-6(10)7-4(8)2-3-5(7)9/h2-3H,1H3

InChI key

LLAZQXZGAVBLRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

For the introduction of the maleimide group in peptides, amino acids and amines[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and some properties of maleimido acids and maleoyl derivatives of peptides.
O Keller et al.
Helvetica chimica acta, 58(2), 531-541 (1975-03-12)
Susanne Kirchhof et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 96, 217-225 (2015-08-09)
Eight-armed PEG was functionalized with furyl and maleimide groups (8armPEG20k-Fur and 8armPEG20k-Mal); degradable hydrogels were obtained by cross-linking via Diels-Alder chemistry. To increase the stability to degradation, the macromonomers were modified by introducing a hydrophobic 6-aminohexanoic acid spacer between PEG

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service