Skip to Content
Merck
All Photos(1)

Key Documents

642878

Sigma-Aldrich

Algal crude protein extract-13C

98 atom % 13C

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€437.00
5 G
€1,470.00

€437.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
€437.00
5 G
€1,470.00

About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.12

€437.00


Check Cart for Availability

Request more information

isotopic purity

98 atom % 13C

Quality Level

form

solid

storage temp.

2-8°C

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karthikeyani Chellappa et al.
Cell metabolism, 34(12), 1947-1959 (2022-12-09)
Nicotinamide adenine dinucleotide (NAD) is an essential redox cofactor in mammals and microbes. Here we use isotope tracing to investigate the precursors supporting NAD synthesis in the gut microbiome of mice. We find that dietary NAD precursors are absorbed in
Samuel Bryson et al.
The ISME journal, 11(12), 2781-2792 (2017-08-12)
Resource availability influences marine microbial community structure, suggesting that population-specific resource partitioning defines discrete niches. Identifying how resources are partitioned among populations, thereby characterizing functional guilds within the communities, remains a challenge for microbial ecologists. We used proteomic stable isotope

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service