Skip to Content
Merck
All Photos(1)

Documents

642541

Sigma-Aldrich

2-Tributylstannylthiazole

97%

Synonym(s):

(Thiazol-2-yl)tributylstannane, 2-(Tributylstannyl)-1,3-thiazole, 2-(Tributylstannyl)thiazole, 2-(Tributyltin)thiazole, Tributyl-2-thiazolylstannane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H29NSSn
CAS Number:
Molecular Weight:
374.17
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.5200 (lit.)

bp

307-309 °C (lit.)

density

1.1900 g/mL at 25 °C (lit.)

SMILES string

CCCC[Sn](CCCC)(CCCC)c1nccs1

InChI

1S/3C4H9.C3H2NS.Sn/c3*1-3-4-2;1-2-5-3-4-1;/h3*1,3-4H2,2H3;1-2H;

InChI key

WUOFQGMXQCSPPV-UHFFFAOYSA-N

Application

Reagent for arylation of thiazole by Stille cross-coupling.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 47, 599-599 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service