Skip to Content
Merck
All Photos(2)

Key Documents

707562

Sigma-Aldrich

5-Chloro-1-aza-5-stanna-bicyclo[3.3.3]undecane

Synonym(s):

Stannatrane

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€184.00
2 G
€669.00

€184.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€184.00
2 G
€669.00

About This Item

Empirical Formula (Hill Notation):
C9H18ClNSn
CAS Number:
Molecular Weight:
294.41
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€184.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

mp

226-230 °C

SMILES string

Cl[Sn]12CCCN(CCC1)CCC2

InChI

1S/C9H18N.ClH.Sn/c1-4-7-10(8-5-2)9-6-3;;/h1-9H2;1H;/q;;+1/p-1

InChI key

BRPCJQRIZVVNKX-UHFFFAOYSA-M

Application

Reactant for:
  • Metal-catalyzed allylic substitution reaction
  • Synthesis of an anti-methicillin-resistant staphylococcus aureus (MRSA) carbapenem via stannatrane mediated Stille coupling

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service