Skip to Content
Merck
All Photos(3)

Key Documents

640484

Sigma-Aldrich

6-Fluoro-2-methylquinoline

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€65.20

€65.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€65.20

About This Item

Empirical Formula (Hill Notation):
C10H8FN
CAS Number:
Molecular Weight:
161.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€65.20


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

49-53 °C (lit.)

functional group

fluoro

SMILES string

Cc1ccc2cc(F)ccc2n1

InChI

1S/C10H8FN/c1-7-2-3-8-6-9(11)4-5-10(8)12-7/h2-6H,1H3

InChI key

GPIARMSVZOEZCV-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service