Skip to Content
Merck
All Photos(1)

Key Documents

640026

Sigma-Aldrich

Poly(pentachlorophenyl methacrylate-co-glycidyl methacrylate)

Glycidyl methacrylate 10 mol %

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€221.00

€221.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
500 MG
€221.00

About This Item

Linear Formula:
(C10H5Cl502)m(C7H10O3)n
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

€221.00


Estimated to ship on21 May 2025


Request a Bulk Order

form

solid

composition

Glycidyl methacrylate, 10 mol %

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service