Skip to Content
Merck
All Photos(2)

Key Documents

433640

Sigma-Aldrich

Poly(ethylene-co-methyl acrylate-co-glycidyl methacrylate)

pellets

Synonym(s):

Ethylene-methyl acrylate-glycidyl methacrylate copolymer, Glycidyl methacrylate-ethylene-methyl acrylate copolymer

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€76.90

€76.90


Estimated to ship on27 March 2025


Request a Bulk Order

Select a Size

Change View
250 G
€76.90

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

€76.90


Estimated to ship on27 March 2025


Request a Bulk Order

form

pellets

Quality Level

melt index

6 g/10 min (190 °C/2.16kg)

composition

glycidyl methacrylate, 8 wt. %
methyl acrylate, 25 wt. %

hardness

70 (Shore A, ASTM D 2240)

mp

39 °C (lit.)

transition temp

softening point <40 °C (Vicat, 1kg load, ASTM D 1525)

density

0.94 g/mL at 25 °C (lit.)

SMILES string

O1C(C1)COC(=O)C(=C)C.O(C)C(=O)C=C.C=C

InChI

1S/C7H10O3.C4H6O2.C2H4/c1-5(2)7(8)10-4-6-3-9-6;1-3-4(5)6-2;1-2/h6H,1,3-4H2,2H3;3H,1H2,2H3;1-2H2

InChI key

CNJPFZOQZWIGIB-UHFFFAOYSA-N

Application

Cross-linkable coatings and adhesion promoter.
Pendant glycidyl functionality available for grafting or cross-linking.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the molecular weight of Poly(ethylene-co-methyl acrylate-co-glycidyl methacrylate)?

    1 answer
    1. The molecular weight of this product is not determined.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service