Skip to Content
Merck
All Photos(1)

Documents

632414

Sigma-Aldrich

Indole-7-carboxaldehyde

97%

Synonym(s):

7-Formylindole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7NO
CAS Number:
Molecular Weight:
145.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

87-91 °C (lit.)

SMILES string

[H]C(=O)c1cccc2cc[nH]c12

InChI

1S/C9H7NO/c11-6-8-3-1-2-7-4-5-10-9(7)8/h1-6,10H

InChI key

XQVZDADGTFJAFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • reactant for preparation of antiandrogens
  • reactant for preparation of antiplatelet agent
  • reactant for preparation of liver X receptor (LXR) agonists
  • reactant for preparation of antagonists of the EP3 receptor for prostaglandin E2
  • reactant for preparation of inhibitors of glycoprotein perforin
  • reactant for preparation of potent antitubulin agents
  • reactant for preparation of potential insulin receptor activators
  • reactant for preparation of potential scaffolds for peptide mimetics
  • reactant for preparation of 5-Hydroxytryptamine1D receptor antagonist
  • reactant for preparation of indolocarbazole kinase inhibitors
Used in an efficient synthesis of fused pyrroloquinolinedicarboxylates from acetylenedicarboxylates and triphenylphosphine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monatshefte fur Chemie / Chemical Monthly, 137, 207-207 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service