Skip to Content
Merck
All Photos(1)

Key Documents

608580

Sigma-Aldrich

Lithium-6Li deuteroxide deuterate

95 atom % 6Li, 98 atom % D

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€537.00

€537.00


Check Cart for Availability


Select a Size

Change View
5 G
€537.00

About This Item

Linear Formula:
6LiOD · D2O
CAS Number:
Molecular Weight:
44.06
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.12

€537.00


Check Cart for Availability

isotopic purity

95 atom % 6Li
98 atom % D

Quality Level

form

solid

mass shift

M+1

storage temp.

room temp

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service