Skip to Content
Merck
All Photos(1)

Documents

590789

Sigma-Aldrich

2,4,5-Trichlorophenol-3,6-d2

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6HD2Cl3O
CAS Number:
Molecular Weight:
199.46
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

bp

248 °C/740 mmHg (lit.)

mp

67-69 °C (lit.)

mass shift

M+2

SMILES string

[2H]c1c(O)c(Cl)c([2H])c(Cl)c1Cl

InChI

1S/C6H3Cl3O/c7-3-1-5(9)6(10)2-4(3)8/h1-2,10H/i1D,2D

InChI key

LHJGJYXLEPZJPM-QDNHWIQGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

271.4 °F - closed cup

Flash Point(C)

133 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service